Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011056.3 | NP_035186.1 | 14 | 1693 | ||
GenBank mRNA | AF536978.1 | - | 14 | 1693 | ||
AK081466.1 | - | 7 | 1066 | |||
AY388962.1 | - | 12 | 1306 | |||
BC132107.1 | - | 13 | 1561 | 40130384 | ||
BC132109.1 | - | 13 | 1561 | 40130386 | ||
DQ665891.1 | - | 7 | 1163 | |||
DQ665892.1 | - | 7 | 949 | |||
DQ665893.1 | - | 8 | 1077 | |||
DQ665894.1 | - | 8 | 921 | |||
DQ665895.1 | - | 7 | 1204 | |||
DQ665896.1 | - | 7 | 993 | |||
EU489880.1 | - | 12 | 1549 | |||
M94541.1 | - | 3 | 316 |