Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289877.1 | NP_001276806.1 | 9 | 1527 | ||
NM_001289878.1 | NP_001276807.1 | 10 | 1576 | |||
NM_011067.3 | NP_035197.2 | 9 | 1527 | |||
GenBank mRNA | AB013605.1 | - | 9 | 1334 | ||
AF050182.1 | - | 9 | 1451 | |||
AJ007377.1 | - | 9 | 1334 | |||
AK036345.1 | - | 9 | 1444 | |||
AK084610.1 | - | 2 | 2495 | |||
AK166514.1 | - | 9 | 1528 | |||
BC141032.1 | - | 9 | 1125 | 9055644 | ||
BC145178.1 | - | 10 | 1174 |