Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 26 | 4271 | ||
GenBank mRNA | AK138724.1 | - | 11 | 1388 | ||
BC096553.1 | - | 25 | 3707 | |||
U52193.1 | - | 27 | 4177 | |||
U55772.1 | - | 25 | 3581 |