Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042502.2 | NP_001035967.1 | 2 | 879 | ||
NM_001042504.2 | NP_001035969.1 | 3 | 370 | |||
NM_001286942.1 | NP_001273871.1 | 4 | 469 | |||
NM_001287048.1 | NP_001273977.1 | 2 | 879 | |||
NM_011098.4 | NP_035228.2 | 4 | 508 | |||
GenBank mRNA | AB006320.1 | - | 2 | 432 | ||
AB006321.1 | - | 4 | 468 | |||
AF048723.1 | - | 3 | 307 | |||
AF048724.1 | - | 2 | 840 | |||
AF201091.1 | - | 2 | 206 | |||
AK034846.1 | - | 1 | 1738 | |||
AK035083.1 | - | 2 | 537 | |||
AM940438.1 | - | 3 | 174 | |||
AM940439.1 | - | 2 | 132 | |||
BC075660.1 | - | 2 | 544 | 30606653 | ||
U70132.1 | - | 1 | 29 | |||
U80010.1 | - | 4 | 443 | |||
U80011.1 | - | 3 | 323 | |||
U80036.1 | - | 3 | 341 |