Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011109.2 | NP_035239.1 | 4 | 1389 | ||
GenBank mRNA | AF169407.1 | - | 4 | 1389 | ||
AF169408.1 | - | 4 | 1389 | |||
AK018005.1 | - | 4 | 1340 | |||
AK157392.1 | - | 4 | 1346 | |||
AK158364.1 | - | 4 | 1316 | |||
BC111806.1 | - | 4 | 1378 | 40039182 |