Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 19, 20 | 2199 | ||
GenBank mRNA | AK028940.1 | - | 19, 20 | 2197 | ||
AK090096.1 | - | 19, 20 | 2200 | |||
AK132704.1 | - | 19, 20 | 2186 | |||
BC060128.1 | - | 18, 19 | 2046 |