Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 1 | 399 | ||
GenBank mRNA | AK134045.1 | - | 1 | 341 | ||
AK148050.1 | - | 1 | 313 | |||
AK148076.1 | - | 1 | 390 | |||
AK164027.1 | - | 1 | 369 | |||
AK182425.1 | - | 1 | 146 | |||
BC006880.1 | - | 1 | 351 | 3596381 | ||
L19558.1 | - | 1 | 406 | |||
U01063.1 | - | 1 | 320 |