Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011132.2 | NP_035262.2 | 2 | 232 | ||
GenBank mRNA | AF123502.1 | - | 2 | 206 | ||
AK141674.1 | - | 2 | 193 | |||
AK165879.1 | - | 2 | 211 | |||
BC048166.1 | - | 2 | 167 | 6410453 | ||
BC063246.1 | - | 2 | 167 | 6410453 | ||
CJ101052.1 | - | 2 | 236 |