Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002713.3 | NP_002704.1 | 6 | 2106 | ||
NM_014110.4 | NP_054829.2 | 7 | 1925 | |||
NM_138558.2 | NP_612568.1 | 7 | 2207 | |||
XM_017001594.1 | XP_016857083.1 | 6 | 1626 | |||
GenBank mRNA | AF061958.1 | - | 7 | 1853 | ||
AF061959.1 | - | 7 | 2008 | |||
AF126488.1 | - | 7 | 1840 | |||
AX775985.1 | - | 7 | 1841 | |||
BC001597.1 | - | 6 | 1733 | |||
BC003405.1 | - | 1 | 566 | |||
BC013360.2 | - | 6 | 1783 | 3633676 | ||
BC093017.1 | - | 7 | 1845 | 30531222 |