Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011165.3 | NP_035295.1 | 2 | 165 | ||
GenBank mRNA | AF011383.1 | - | 2 | 166 | ||
AF015562.1 | - | 2 | 165 | |||
AK141285.1 | - | 2 | 177 | |||
AK164009.1 | - | 2 | 177 | |||
BC052218.1 | - | 2 | 166 | 30017285 |