Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 22, 23 | 3447 | ||
NM_005424.4 | NP_005415.1 | 22, 23 | 3530 | |||
XM_005271163.1 | XP_005271220.1 | 21, 22 | 3344 | |||
XM_006710869.1 | XP_006710932.1 | 22, 23 | 3664 | |||
GenBank mRNA | AF217976.1 | - | 12, 13 | 1614 | ||
AK225560.1 | - | 22, 23 | 3189 | |||
AK296735.1 | - | 17, 18 | 2506 | |||
AK300399.1 | - | 22, 23 | 3337 | |||
BC038239.1 | - | 22, 23 | 3432 | 5767075 | ||
X60957.1 | - | 22, 23 | 3389 |