Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011217.2 | NP_035347.1 | 2 | 544 | ||
GenBank mRNA | AK028054.1 | - | 2 | 545 | ||
AK051719.1 | - | 2 | 621 | |||
BC119230.1 | - | 2 | 477 | 8733979 | ||
BC120547.1 | - | 2 | 477 | 40129470 | ||
D31898.1 | - | 2 | 507 |