Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163478.1 | NP_001156950.1 | 4 | 392 | ||
NM_001163479.1 | NP_001156951.1 | 2 | 330 | |||
NM_011231.2 | NP_035361.2 | 3 | 530 | |||
GenBank mRNA | AF231926.1 | - | 3 | 386 | ||
AK010690.1 | - | 2 | 82 | |||
AK160162.1 | - | 4 | 379 | |||
BC003948.1 | - | 3 | 373 | |||
BC021335.1 | - | 3 | 376 | |||
BC057661.1 | - | 2 | 962 | |||
BC132473.1 | - | 3 | 408 | 40130750 | ||
BC138547.1 | - | 3 | 408 | 8861567 | ||
BC145323.1 | - | 3 | 408 | |||
BC145324.1 | - | 3 | 408 | |||
BC145325.1 | - | 3 | 408 | |||
BC145326.1 | - | 3 | 368 | |||
BG076913.2 | - | 2 | 99 | |||
CF732295.1 | - | 3 | 365 | |||
CN456589.1 | - | 2 | 239 | |||
CV562676.1 | - | 2 | 176 | |||
CX205055.1 | - | 4 | 392 | |||
U12922.1 | - | 3 | 386 |