Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164767.1 | NP_001158239.1 | 3 | 789 | ||
GenBank mRNA | AF060570.1 | - | 2 | 775 |