Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290561.1 | NP_001277490.1 | 3 | 469 | ||
NM_001290562.1 | NP_001277491.1 | 3 | 469 | |||
NM_011123.3 | NP_035253.1 | 3 | 469 | |||
GenBank mRNA | AK077564.1 | - | 3 | 427 | ||
AK134554.1 | - | 3 | 460 | |||
AK160872.1 | - | 3 | 464 | |||
BC027010.1 | - | 3 | 379 | 5364736 | ||
M14674.1 | - | 2 | 303 | |||
M15442.1 | - | 3 | 448 | |||
M16472.1 | - | 2 | 303 | |||
M20752.1 | - | 2 | 303 |