Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 4 | 452 | ||
GenBank mRNA | AK148330.1 | - | 4 | 457 | ||
AK149730.1 | - | 4 | 383 | |||
AK190857.1 | - | 1 | 116 | |||
BC003782.1 | - | 4 | 386 | 3491360 | ||
CT010317.1 | - | 3 | 316 | |||
U37226.1 | - | 4 | 382 |