Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163554.1 | NP_001157026.1 | 11 | 1021 | ||
NM_001163555.1 | NP_001157027.1 | 11 | 1087 | |||
NM_001163556.1 | NP_001157028.1 | 11 | 1021 | |||
NM_011138.2 | NP_035268.2 | 11 | 1069 | |||
GenBank mRNA | AK156322.1 | - | 11 | 1034 | ||
AY746973.1 | - | 11 | 1049 | |||
AY746974.1 | - | 11 | 1067 | |||
BC050258.1 | - | 11 | 1034 | |||
BC104488.1 | - | 11 | 1021 | 40046281 | ||
BC105647.1 | - | 11 | 1021 | 40046279 | ||
BC105920.1 | - | 11 | 1087 | 40046289 | ||
BC105921.1 | - | 11 | 1021 | 40046286 | ||
X53654.1 | - | 1229 | ||||
X57936.1 | - | 11 | 1021 | |||
X57937.1 | - | 11 | 1069 | |||
X57938.1 | - | 11 | 1087 | |||
X57939.1 | - | 11 | 1021 | |||
X57940.1 | - | 11 | 1021 | |||
X57941.1 | - | 10 | 904 |