Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136540.1 | NP_001130012.1 | 6 | 1352 | ||
NM_001136541.1 | NP_001130013.1 | 5 | 1259 | |||
NM_003661.3 | NP_003652.2 | 6 | 1313 | |||
NM_145343.2 | NP_663318.1 | 7 | 1467 | |||
XM_005261796.3 | XP_005261853.1 | 5 | 1238 | |||
XM_011530478.2 | XP_011528780.1 | 6 | 1385 | |||
GenBank mRNA | AF019225.2 | - | 6 | 1312 | ||
AF305224.1 | - | 6 | 1252 | |||
AF323540.1 | - | 7 | 1466 | |||
AK298053.1 | - | 3 | 1043 | |||
AK300454.1 | - | 5 | 1228 | |||
AK303919.1 | - | 6 | 1286 | |||
AK309143.1 | - | 7 | 1498 | |||
AK313752.1 | - | 7 | 1406 | |||
BC017331.1 | - | 6 | 1243 | |||
BC112943.1 | - | 6 | 1268 | 4512933 | ||
BC113867.1 | - | 6 | 1243 | |||
BC114475.1 | - | 6 | 1243 | |||
BC127186.1 | - | 6 | 1243 | 40034503 | ||
BC141823.1 | - | 6 | 1268 | 40148156 | ||
BC142720.1 | - | 6 | 1268 | |||
BC143038.1 | - | 6 | 1243 | 40034505 | ||
BC143039.1 | - | 6 | 1243 | 40034506 |