Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112701.1 | NP_001106171.1 | 6 | 676 | ||
NM_011181.3 | NP_035311.1 | 6 | 676 | |||
GenBank mRNA | AB013466.1 | - | 6 | 611 | ||
AB013467.1 | - | 5 | 1179 | |||
AF079971.1 | - | 6 | 645 | |||
AK045451.1 | - | 6 | 645 | |||
AK078067.1 | - | 6 | 672 | |||
AK143829.1 | - | 6 | 618 | |||
AK168979.1 | - | 6 | 616 | |||
AK172050.1 | - | 6 | 632 | |||
BC004662.1 | - | 6 | 590 | 3498490 | ||
BC088833.1 | - | 6 | 650 | 5293465 |