Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011192.3 | NP_035322.1 | 5, 6 | 486 | ||
GenBank mRNA | AK140597.1 | - | 5, 6 | 507 | ||
AK150939.1 | - | 5, 6 | 506 | |||
AK156079.1 | - | 5, 6 | 456 | |||
AK171348.1 | - | 5, 6 | 502 | |||
AK178475.1 | - | 4, 5 | 205 | |||
AK183517.1 | - | 3, 4 | 230 | |||
AK184446.1 | - | 3, 4 | 230 | |||
AK198462.1 | - | 3, 4 | 230 | |||
AK210357.1 | - | 3, 4 | 226 | |||
BC015255.1 | - | 5, 6 | 486 | 4221225 | ||
BC139818.1 | - | 5, 6 | 434 | 40141268 | ||
CT010198.1 | - | 5, 6 | 290 | |||
D87911.1 | - | 5, 6 | 409 | |||
U60330.1 | - | 5, 6 | 290 |