Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011206.2 | NP_035336.2 | 3 | 326 | ||
GenBank mRNA | AK144353.1 | - | 3 | 308 | ||
BC008512.1 | - | 3 | 296 | 3591418 | ||
U35124.1 | - | 3 | 300 | |||
U49853.1 | - | 3 | 266 | |||
U52523.1 | - | 3 | 314 |