Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011224.1 | NP_035354.1 | 6 | 802 | ||
GenBank mRNA | AF124787.1 | - | 6 | 802 | ||
BC012961.1 | - | 6 | 808 | 3989941 |