Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163478.1 | NP_001156950.1 | 6 | 536 | ||
NM_001163479.1 | NP_001156951.1 | 4 | 474 | |||
NM_011231.2 | NP_035361.2 | 5 | 674 | |||
GenBank mRNA | AF231926.1 | - | 6 | 628 | ||
AK010690.1 | - | 4 | 226 | |||
AK159902.1 | - | 7 | 982 | |||
AK160162.1 | - | 6 | 523 | |||
BC003948.1 | - | 6 | 615 | |||
BC021335.1 | - | 5 | 520 | |||
BC057661.1 | - | 5 | 1204 | |||
BC132473.1 | - | 5 | 552 | 40130750 | ||
BC138547.1 | - | 5 | 552 | 8861567 | ||
BC145323.1 | - | 6 | 650 | |||
BC145324.1 | - | 5 | 552 | |||
BC145325.1 | - | 6 | 650 | |||
BC145326.1 | - | 6 | 610 | |||
BG076913.2 | - | 4 | 243 | |||
CF732295.1 | - | 6 | 607 | |||
CN456589.1 | - | 4 | 383 | |||
CV562676.1 | - | 5 | 418 | |||
U12922.1 | - | 5 | 530 |