Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 11 | 1809 | ||
NM_014491.3 | NP_055306.1 | 11 | 1812 | |||
NM_148898.3 | NP_683696.2 | 12 | 1887 | |||
NM_148900.3 | NP_683698.2 | 12 | 1863 | |||
NR_033767.1 | 12 | 1870 | ||||
XM_017012801.1 | XP_016868290.1 | 14 | 2493 | |||
GenBank mRNA | AF086040.1 | - | 1 | 202 | ||
AF337817.1 | - | 11 | 1588 | |||
AK296957.1 | - | 12 | 1743 | |||
AY144615.1 | - | 12 | 1882 | |||
BC126104.1 | - | 12 | 1544 | 8991820 | ||
BC143866.1 | - | 12 | 1544 | 9052382 | ||
BC143867.1 | - | 11 | 1483 | 9052383 | ||
CR749236.1 | - | 18 | 2193 | |||
KJ534839.1 | - | 11 | 1513 | |||
KJ535079.1 | - | 10 | 1651 | |||
KJ535086.1 | - | 10 | 1437 |