Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011239.2 | NP_035369.2 | 3 | 390 | ||
GenBank mRNA | AK002989.1 | - | 3 | 356 | ||
AK088781.1 | - | 3 | 378 | |||
AK151167.1 | - | 3 | 368 | |||
AK153074.1 | - | 3 | 372 | |||
AK166642.1 | - | 3 | 392 | |||
AK167053.1 | - | 3 | 374 | |||
BC061140.1 | - | 3 | 409 | 6775220 | ||
L25255.1 | - | 3 | 263 | |||
X56045.1 | - | 3 | 395 | |||
X56046.1 | - | 3 | 395 |