Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011254.5 | NP_035384.1 | 2 | 2242 | ||
GenBank mRNA | AK011641.1 | - | 2 | 390 | ||
BC018254.1 | - | 2 | 369 | 3498107 | ||
BC091751.1 | - | 2 | 386 | 4022443 | ||
X60367.1 | - | 2 | 2241 |