Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 5, 6 | 632 | ||
GenBank mRNA | AK148330.1 | - | 5, 6 | 637 | ||
AK149730.1 | - | 5, 6 | 563 | |||
AK190857.1 | - | 2, 3 | 296 | |||
BC003782.1 | - | 5, 6 | 566 | 3491360 | ||
CT010317.1 | - | 4, 5 | 496 | |||
U28960.1 | - | 4, 5 | 490 | |||
U37226.1 | - | 5, 6 | 562 |