Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152323.1 | NP_689536.1 | 5 | 446 | ||
XM_005268646.3 | XP_005268703.1 | 6 | 724 | |||
XM_006719239.3 | XP_006719302.1 | 6 | 690 | |||
GenBank mRNA | AF518404.1 | - | 6 | 579 | ||
BC032317.1 | - | 5 | 446 | 5222618 |