Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152380.2 | NP_689593.2 | 8 | 1912 | ||
XM_005271161.3 | XP_005271218.1 | 9 | 2467 | |||
XM_005271162.1 | XP_005271219.1 | 8 | 2368 | |||
XM_011542052.2 | XP_011540354.1 | 5 | 1413 | |||
GenBank mRNA | AK096396.1 | - | 4 | 1199 | ||
AK127536.1 | - | 8 | 1912 | |||
BC122553.1 | - | 8 | 1730 | 40077247 | ||
BX537778.1 | - | 3 | 1098 |