Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112701.1 | NP_001106171.1 | 12 | 2453 | ||
NM_011181.3 | NP_035311.1 | 13 | 2456 | |||
GenBank mRNA | AK045451.1 | - | 13 | 2425 | ||
BY470655.1 | - | 1 | 359 |