Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011189.1 | NP_035319.1 | 7 | 518 | ||
GenBank mRNA | AK008514.1 | - | 7 | 493 | ||
AK019390.1 | - | 7 | 546 | |||
AK150708.1 | - | 7 | 562 | |||
AK150975.1 | - | 7 | 561 | |||
AK182144.1 | - | 3 | 210 | |||
BC035549.1 | - | 2 | 1182 | |||
BC088981.1 | - | 7 | 538 | 5689048 | ||
D87909.1 | - | 7 | 518 | |||
U60328.1 | - | 7 | 437 |