Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164401.1 | NP_001157873.1 | 20 | 3306 | ||
NM_001164402.1 | NP_001157874.1 | 10 | 936 | |||
NM_001164403.1 | NP_001157875.1 | 9 | 852 | |||
NM_011216.3 | NP_035346.3 | 21 | 3390 | |||
GenBank mRNA | AF135166.1 | - | 20 | 3271 | ||
AF295638.1 | - | 21 | 3003 | |||
BC051976.1 | - | 10 | 849 | |||
BC052743.1 | - | 21 | 3224 | 6834312 | ||
BC063258.1 | - | 11 | 1045 | |||
BC079649.1 | - | 852 | ||||
U37465.1 | - | 10 | 920 | |||
U37466.1 | - | 9 | 836 |