Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044371.2 | NP_001037836.1 | 17 | 2207 | ||
NM_001283011.1 | NP_001269940.1 | 18 | 2412 | |||
NM_011233.3 | NP_035363.2 | 17 | 2335 | |||
GenBank mRNA | AF085737.1 | - | 17 | 2258 | ||
AJ011923.1 | - | 16 | 2105 | |||
AK134277.1 | - | 17 | 2162 | |||
AK162016.1 | - | 17 | 2195 | |||
AK167358.1 | - | 18 | 2355 | |||
AK201395.1 | - | 1 | 84 | |||
BC066855.1 | - | 17 | 2325 | 30473984 |