Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042727.2 | NP_001036192.1 | 4 | 747 | ||
NM_011244.4 | NP_035374.3 | 6 | 985 | |||
GenBank mRNA | AK148064.1 | - | 6 | 982 | ||
AK155911.1 | - | 4 | 600 | |||
BC012923.1 | - | 6 | 911 | 3154037 | ||
BC013709.1 | - | 4 | 539 | 3989967 | ||
M34475.1 | - | 4 | 523 | |||
M34476.1 | - | 6 | 742 | |||
X15848.1 | - | 6 | 805 |