Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011246.2 | NP_035376.1 | 4, 5 | 565 | ||
GenBank mRNA | AF106070.1 | - | 4, 5 | 409 | ||
AK013548.1 | - | 4, 5 | 611 | |||
AK134005.1 | - | 4, 5 | 579 | |||
AK141524.1 | - | 4, 5 | 690 | |||
BC057120.1 | - | 4, 5 | 565 | 6831137 | ||
BC057341.1 | - | 4, 5 | 565 | 6831137 |