Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011274.5 | NP_035404.4 | 4 | 670 | ||
GenBank mRNA | AF091096.1 | - | 2 | 192 | ||
AK133383.1 | - | 4 | 345 | |||
AK155994.1 | - | 4 | 644 | |||
AK160558.1 | - | 4 | 610 | |||
AK166566.1 | - | 4 | 661 | |||
AK170929.1 | - | 4 | 665 | |||
BC023029.1 | - | 4 | 330 | 5374731 |