Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011292.2 | NP_035422.1 | 3 | 172 | ||
GenBank mRNA | AF260271.1 | - | 2 | 142 | ||
AK012331.1 | - | 3 | 142 | |||
AK012445.1 | - | 3 | 142 | |||
AK017388.1 | - | 3 | 142 | |||
AK017427.1 | - | 3 | 142 | |||
AK050296.1 | - | 2 | 231 | |||
AK053431.1 | - | 2 | 525 | |||
AK084278.1 | - | 3 | 143 | |||
AK088166.1 | - | 3 | 142 | |||
AK153562.1 | - | 3 | 142 | |||
AK159980.1 | - | 3 | 142 | |||
AK168326.1 | - | 3 | 140 | |||
AK169217.1 | - | 3 | 140 | |||
BC013165.1 | - | 2 | 132 | 2655358 | ||
BC081435.1 | - | 2 | 118 | 6824107 | ||
BC083166.1 | - | 2 | 123 | 6397429 | ||
BC083329.1 | - | 2 | 121 | 6824346 | ||
BC086937.1 | - | 3 | 139 | 6787671 | ||
BC089319.1 | - | 2 | 121 | 6823996 | ||
BC093526.1 | - | 2 | 121 | 5150493 | ||
BY072215.1 | - | 3 | 175 | |||
U17331.1 | - | 1 | 78 | |||
U17332.1 | - | 1 | 78 |