Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011310.2 | NP_035440.1 | 3 | 387 | ||
GenBank mRNA | AF004941.1 | - | 3 | 387 | ||
AF087470.1 | - | 3 | 362 | |||
AK133717.1 | - | 3 | 389 | |||
AK161105.1 | - | 1 | 2134 | |||
BC099893.1 | - | 3 | 384 | 40043723 | ||
BC100690.1 | - | 3 | 384 | 40043727 | ||
BC100696.1 | - | 3 | 384 | 40043722 | ||
BC100697.1 | - | 3 | 384 | 40043719 |