Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI226497.1 | - | 2 | 375 | ||
AK014418.1 | - | 2 | 345 | |||
AK157860.1 | - | 2 | 210 | |||
AK163400.1 | - | 2 | 330 | |||
AK167113.1 | - | 2 | 259 | |||
BC005715.1 | - | 3 | 481 | |||
BC068152.1 | - | 2 | 585 | 30534982 | ||
D50461.1 | - | 2 | 588 | |||
U45977.1 | - | 2 | 493 | |||
U45978.1 | - | 2 | 361 |