Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153442.3 | NP_703143.1 | 3 | 3583 | ||
GenBank mRNA | AJ505757.1 | - | 3 | 3582 |
Set Membership: |
G-protein coupled receptor |