Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 2 | 1336 | ||
NM_001289538.1 | NP_001276467.1 | 2 | 1336 | |||
NM_001289539.1 | NP_001276468.1 | 2 | 959 | |||
NM_001289540.1 | NP_001276469.1 | 3 | 1096 | |||
NM_001289541.1 | NP_001276470.1 | 2 | 959 | |||
NM_001289542.1 | NP_001276471.1 | 2 | 1336 | |||
NM_011363.3 | NP_035493.2 | 2 | 1336 | |||
GenBank mRNA | AF020526.1 | - | 2 | 1030 | ||
AF074329.1 | - | 1 | 741 | |||
AF380422.1 | - | 2 | 1030 | |||
AF421138.1 | - | 2 | 1030 | |||
AF421139.1 | - | 2 | 1030 | |||
AK148085.1 | - | 3 | 1063 | |||
AK154655.1 | - | 2 | 1270 | |||
AK168439.1 | - | 2 | 931 | |||
AK170444.1 | - | 2 | 1310 | |||
AK171328.1 | - | 2 | 1339 | |||
AK173145.1 | - | 1 | 483 | |||
AK187869.1 | - | 1 | 175 | |||
AK191125.1 | - | 1 | 175 | |||
BC011422.1 | - | 1 | 798 | 3156603 | ||
BC051978.1 | - | 2 | 879 | 6401205 |