Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011374.2 | NP_035504.2 | 1, 2 | 925 | ||
GenBank mRNA | AK046188.1 | - | 1, 2 | 865 | ||
AK052444.1 | - | 1, 2 | 576 | |||
BC024821.1 | - | 1, 2 | 925 | 5357151 | ||
EF101558.1 | - | 1, 2 | 37 | |||
L38677.1 | - | 1, 2 | 549 | |||
X84235.1 | - | 1, 2 | 272 |