Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011297.2 | NP_035427.2 | 4 | 407 | ||
NM_207634.1 | NP_997517.1 | 4 | 407 | |||
NM_207635.1 | NP_997518.1 | 4 | 407 | |||
GenBank mRNA | AK002495.1 | - | 4 | 371 | ||
AK002986.1 | - | 770 | ||||
AK005358.1 | - | 4 | 371 | |||
AK007776.1 | - | 4 | 371 | |||
AK008386.1 | - | 4 | 371 | |||
AK009048.1 | - | 4 | 371 | |||
AK010856.1 | - | 4 | 371 | |||
AK012537.1 | - | 4 | 358 | |||
AK018704.1 | - | 4 | 371 | |||
AK049387.1 | - | 367 | ||||
AK052480.1 | - | 3 | 1639 | |||
AK135247.1 | - | 4 | 360 | |||
AK151111.1 | - | 4 | 369 | |||
AK167874.1 | - | 3 | 342 | |||
BC058817.1 | - | 4 | 367 | 30058168 | ||
BC081457.1 | - | 4 | 353 | 5712639 | ||
BC086882.1 | - | 4 | 370 | 30120790 | ||
BC091748.1 | - | 4 | 351 | 2812455 | ||
BC092287.1 | - | 3 | 338 | |||
X60289.1 | - | 4 | 362 |