Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011304.3 | NP_035434.1 | 5, 6 | 605 | ||
GenBank mRNA | AB013912.1 | - | 5, 6 | 436 | ||
AK135404.1 | - | 5, 6 | 419 | |||
AK144156.1 | - | 5, 6 | 427 | |||
AK159112.1 | - | 5, 6 | 607 | |||
BC072585.1 | - | 1104 | ||||
BC115809.1 | - | 5, 6 | 423 | 40105495 | ||
BC116693.1 | - | 5, 6 | 422 | 40105493 |