Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011347.2 | NP_035477.1 | 3 | 316 | ||
GenBank mRNA | BC109158.1 | - | 2 | 202 | 40047544 | |
BC109159.1 | - | 2 | 202 | 40047547 | ||
M72332.1 | - | 3 | 316 | |||
M87861.1 | - | 2 | 202 |