Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011355.1 | NP_035485.1 | 4, 5 | 659 | ||
GenBank mRNA | AK142246.1 | - | 4, 5 | 676 | ||
AK153524.1 | - | 4, 5 | 690 | |||
AK154945.1 | - | 4, 5 | 711 | |||
AK155085.1 | - | 4, 5 | 695 | |||
BC003815.1 | - | 4, 5 | 577 | 3600260 | ||
L03215.1 | - | 4, 5 | 556 | |||
M32370.1 | - | 4, 5 | 639 | |||
M38252.1 | - | 4, 5 | 659 | |||
X17463.1 | - | 4, 5 | 1347 |