Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011364.3 | NP_035494.1 | 3, 4 | 466 | ||
GenBank mRNA | AF072903.1 | - | 3, 4 | 513 | ||
AF097632.1 | - | 3, 4 | 412 | |||
AF097633.1 | - | 3, 4 | 405 | |||
AK040076.1 | - | 3, 4 | 467 | |||
AK042476.1 | - | 295 | ||||
BC028467.2 | - | 1, 2 | 1057 | |||
BC065390.1 | - | 3, 4 | 436 | |||
BC147367.1 | - | 3, 4 | 376 | 9056424 | ||
BC147368.1 | - | 3, 4 | 376 | 9056435 | ||
BC147807.1 | - | 3, 4 | 239 | 9087524 | ||
BC147811.1 | - | 3, 4 | 239 | 9087528 | ||
KF811030.1 | - | 4, 5 | 405 |