Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011374.2 | NP_035504.2 | 5 | 1515 | ||
GenBank mRNA | AK042207.1 | - | 5 | 1371 | ||
AK046188.1 | - | 5 | 1455 | |||
AK052444.1 | - | 5 | 1166 | |||
BC024821.1 | - | 5 | 1515 | 5357151 | ||
L38677.1 | - | 5 | 1139 |