Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011384.4 | NP_035514.1 | 2 | 936 | ||
GenBank mRNA | AF135267.1 | - | 2 | 987 | ||
AK017544.1 | - | 2 | 997 | |||
AK029309.1 | - | 2 | 937 | |||
BC138838.1 | - | 2 | 909 | 8861860 | ||
BC138839.1 | - | 2 | 909 | 8861861 |