Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011258.2 | NP_035388.2 | 2, 3 | 244 | ||
GenBank mRNA | AK132028.1 | - | 2, 3 | 224 | ||
AK136937.1 | - | 2, 3 | 223 | |||
AK146164.1 | - | 2, 3 | 239 | |||
AK146314.1 | - | 2, 3 | 239 | |||
BC085173.1 | - | 2, 3 | 221 | 30549691 | ||
BC085174.1 | - | 2, 3 | 213 | |||
BY714128.1 | - | 2, 3 | 272 | |||
U01222.1 | - | 2, 3 | 212 | |||
U07157.1 | - | 2, 3 | 168 | |||
U15037.1 | - | 2, 3 | 245 | |||
U36441.1 | - | 2, 3 | 221 | |||
X72711.1 | - | 2, 3 | 210 |